Key Documents
Z508179
Ace small soap bubble meter
Tube only
About This Item
UNSPSC Code:
41121800
Pricing and availability is not currently available.
General description
For measuring gas flow directly. Graduated to 100mL in 0.2mL increments, with a range of 6mL to 3,000mL/min. #15 Ace-Thred at top with nylon bushing and PTFE ferrule allows positioning of the bubble breaker tube. The side stopcock near the top can be connected to a manometer or drain hose.
Choose from one of the most recent versions:
Certificates of Analysis (COA)