Skip to Content
Merck
All Photos(1)

Documents

W343609

Sigma-Aldrich

1-Methyl-3-methoxy-4-isopropylbenzene

98%

Synonym(s):

2-Isopropyl-5-methylanisole, O-Methylthymol, NSC 404221, Thymol methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
FEMA Number:
3436
EC Number:
Council of Europe no.:
11245
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.043
NACRES:
NA.21

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 117

Assay

98%

refractive index

n20/D 1.5063 (lit.)

bp

214-216 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; musty; earthy; herbaceous

SMILES string

COc1cc(C)ccc1C(C)C

InChI

1S/C11H16O/c1-8(2)10-6-5-9(3)7-11(10)12-4/h5-8H,1-4H3

InChI key

LSQXNMXDFRRDSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Coffee, french fried potato.

Biochem/physiol Actions

Taste at 1.0-5.0 ppm

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 102-102 (2003)
Yongpeng Ma et al.
Journal of ethnopharmacology, 231, 386-393 (2018-11-25)
A folk herb Adenosma buchneroides found in the previous ethnobotanical investigation plays an important role as an insect repellent among the Aini people in southwest of China, but the active compounds responsible for repellent activity of the plant have not
Hatice Büşra Konuk et al.
Folia microbiologica, 65(4), 775-783 (2020-03-21)
Terpenoids, one of the major components of essential oils, are known to exert potent antifungal activity against yeast Saccharomyces cerevisiae. They have been the subject of a considerable number of investigations that uncovered extensive pharmacological properties, including antifungal and antibacterial
Elena Canellas et al.
Journal of mass spectrometry : JMS, 49(11), 1181-1190 (2014-11-15)
Pressure-sensitive adhesives (PSA) are used to manufacture labels that are applied directly on the food. These adhesives could contain not only intentionally added compounds (IAS) to the adhesive formula but also non-intentionally added substances (NIAS), due to the impurities from
E Canellas et al.
Food chemistry, 197(Pt A), 24-29 (2015-12-01)
Most fresh sausages are sold with a self-stick adhesive label stuck directly on it. Because of that, the substances in the adhesive could migrate into the fresh sausage. In this work, the multiple headspace-solid-phase microextraction technique has been optimized to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service