Skip to Content
Merck
All Photos(1)

Key Documents

CDS014278

Sigma-Aldrich

N-benzyl-9H-purin-6-amine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
Molecular Weight:
225.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

C(Nc1ncnc2[nH]cnc12)c3ccccc3

InChI

1S/C12H11N5/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11/h1-5,7-8H,6H2,(H2,13,14,15,16,17)

InChI key

NWBJYWHLCVSVIJ-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrizia D'Amelio et al.
Bone, 76, 177-184 (2015-04-02)
We investigated the effect of bone turnover on glucose homeostasis, fat distribution and adipokine production during anabolic treatment with PTH. This is a parallel, randomized controlled, open label, trial. The randomization was done by computer generated tables to allocate treatments.
Raúl A Almeida et al.
Veterinary research, 46, 133-133 (2015-11-20)
In the present study, the effect of anti-recombinant Streptococcus uberis adhesion molecule (SUAM) antibodies against S. uberis intramammary infections (IMI) was evaluated using a passive protection model. Mammary quarters of healthy cows were infused with S. uberis UT888 opsonized with
Alexandar Tzankov et al.
Journal of hematology & oncology, 8, 70-70 (2015-06-14)
The prognostic role of tumor-related parameters in diffuse large B cell lymphoma (DLBCL) is a matter of controversy. We investigated the prognostic value of phenotypic and genotypic profiles in DLBCL in clinical trial (NCT00544219) patients homogenously treated with six cycles
Anke Doyon et al.
PloS one, 10(2), e0113482-e0113482 (2015-02-07)
The extent and relevance of altered bone metabolism for statural growth in children with chronic kidney disease is controversial. We analyzed the impact of renal dysfunction and recombinant growth hormone therapy on a panel of serum markers of bone metabolism
Paul J Kostenuik et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 30(4), 657-669 (2014-11-06)
Postmenopausal osteoporosis is a chronic disease wherein increased bone remodeling reduces bone mass and bone strength. Antiresorptive agents including bisphosphonates are commonly used to mitigate bone loss and fracture risk. Osteoclast inhibition via denosumab (DMAb), a RANKL inhibitor, is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service