Skip to Content
Merck
All Photos(1)

Documents

932868

Sigma-Aldrich

Tweezers

Style AA, ESD Rubber Coated Boley

Synonym(s):

Tweezer, Gripper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

Safe ergonomic cushion grips (resistivity 10E9 Ohm) for handling ESD sensitive components or small static items. Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
OAL: 130 mm. Material: Anti-Acid/Anti-MagSS. Tips: Straight, sharp, fine.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service