Skip to Content
Merck
All Photos(1)

Key Documents

921556

Sigma-Aldrich

Graphene oxide

organic solvent dispersible

Synonym(s):

Graphene oxide, Graphene oxide for non-aqueous solvent dispersions, Graphene oxide for organic solvent dispersions

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxHyOz
CAS Number:
UNSPSC Code:
12352119
NACRES:
NA.23

Quality Level

form

solid

solubility

2-propanol: soluble 2 mg/mL (IPA)
DMSO: soluble 2 mg/mL (dimethyl sulfoxide)
NMP: soluble 2 mg/mL (1-methyl-2-pyrrolidinone)
THF: soluble 2 mg/mL (tetrahydrofuran )
dichloromethane: soluble 2 mg/mL (DCM)
ethyl acetate: soluble 2 mg/mL (EtOAc)
soluble (dispersible in organic solvents)

General description

This graphene oxide product has been formulated to make the graphene oxide dispersible in many anhydrous organic solvents including dichloromethane (DCM), dimethyl sulfoxide (DMSO), 2-propanol (IPA), tetrahydrofuran (THF), and 1-methyl-2-pyrrolidinone (NMP).

Application

Our reformulated non-covalently modified graphene oxide allows you to expore the unique properties of graphene oxide in new ways that were not previously possible. You can use our hydrophobic graphene oxide to make composites with polymers and other nanomaterials that are incompatiable with or insoluble in water. Additionaly, graphene oxide may be reduced to give electrically conductive composites that have been used in applications such as fuel cells, photocatalysis, supercapacitors, lithium-ion batteries, sodium-ion batteries, and lithium sulfur batteries.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graphene/Polyaniline Nanofiber Composites as Supercapacitor Electrodes.
Zhang K, et al.
Chemistry of Materials, 22(4), 1392-1401 (2010)
Yongming Sun et al.
ACS nano, 5(9), 7100-7107 (2011-08-10)
Self-assembled hierarchical MoO(2)/graphene nanoarchitectures have been fabricated on a large scale through a facile solution-phase process and subsequent reduction of the Mo-precursor/graphene composite. The as-formed MoO(2)/graphene nanohybrid as an anode material for lithium-ion batteries exhibits not only a highly reversible
Lamuel David et al.
ACS nano, 8(2), 1759-1770 (2014-01-23)
We study the synthesis and electrochemical and mechanical performance of layered free-standing papers composed of acid-exfoliated few-layer molybdenum disulfide (MoS2) and reduced graphene oxide (rGO) flakes for use as a self-standing flexible electrode in sodium-ion batteries. Synthesis was achieved through
Liwen Ji et al.
Journal of the American Chemical Society, 133(46), 18522-18525 (2011-10-25)
The loss of sulfur cathode material as a result of polysulfide dissolution causes significant capacity fading in rechargeable lithium/sulfur cells. Here, we use a chemical approach to immobilize sulfur and lithium polysulfides via the reactive functional groups on graphene oxide.
Sheng Chen et al.
ACS nano, 4(5), 2822-2830 (2010-04-14)
A composite of graphene oxide supported by needle-like MnO(2) nanocrystals (GO-MnO(2) nanocomposites) has been fabricated through a simple soft chemical route in a water-isopropyl alcohol system. The formation mechanism of these intriguing nanocomposites investigated by transmission electron microscopy and Raman

Articles

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service