Skip to Content
Merck
All Photos(1)

Key Documents

906840

Sigma-Aldrich

Poly(L-lactide-co-caprolactone)

lactide:caprolactone 90:10, viscosity 1.7 dL/g 

Synonym(s):

PCL-PLLA, PLLA-PCL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C6H10O2)m(C6H8O4)nCH3
UNSPSC Code:
12162002

form

chunks

feed ratio

lactide:caprolactone 90:10

color

white to off-white

viscosity

1.7 dL/g

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(L-lactide-co-caprolactone) (PCL-PLLA) is a biodegradable polymer which can be synthesized by bulk ring-opening copolymerization of L-lactide and ε-caprolactone. Its properties include flexibility, biocompatibility and controlled degradation process.

Application

PCL-PLLA can be used for a variety of applications such as tissue engineering, bone fixation devices and surgical sutures.
Poly(L-lactide-co-caprolactone) (PLLA-PCL) is a biodegradable copolymer. poly(L-lactide) exhibits a high modulus and has been often copolymerized with PCL to improve the mechanical properties of PCL, and increase degradation rate of the resulting polymer. PLLA-PCL has been used in tissue engineering, 3D bioprinting and drug delivery research applications.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of a bilayer tubular scaffold based on electrospun poly (L-lactide-co-caprolactone)/collagen fibers and yarns for tracheal tissue engineering
Wu T, et al.
Journal of materials chemistry. B, 5(1), 139-150 (2017)
Preclinical animal study and human clinical trial data of co-electrospun poly (L-lactide-co-caprolactone) and fibrinogen mesh for anterior pelvic floor reconstruction
Wu X, et al.
International journal of nanomedicine, 11(6), 389-389 (2016)
Preparation of a poly (L-lactide-co-caprolactone) copolymer using a novel tin (II) alkoxide initiator and its fiber processing for potential use as an absorbable monofilament surgical suture
Ruengdechawiwat S, et al.
International Journal of Polymeric Materials and Polymeric Biomaterials, 65(6), 277-284 (2016)
Comparison of plasma and chemical modifications of poly-L-lactide-co-caprolactone scaffolds for heparin conjugation
Hsieh Y, et al.
Bio-medical materials and engineering, 12(6), 065004-065004 (2017)
Fabrication and characterization of vitamin B5 loaded poly (l-lactide-co-caprolactone)/silk fiber aligned electrospun nanofibers for schwann cell proliferation
Bhutto MA, et al.
Colloids and Surfaces. B, Biointerfaces, 144(6), 108-117 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service