Skip to Content
Merck
All Photos(1)

Key Documents

89184

Sigma-Aldrich

Alloc protecting group introducing reagent, polymer-bound

extent of labeling: ~1.0 mmol/g loading

Synonym(s):

1-(Alloc-oxy)benzotriazole-6-carboxamidomethyl-polystyrene, Alloc-OBt, polymer-bound, Benzotriazol-1-yl allyl carbonate, polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

crystals

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: amine reactive

extent of labeling

~1.0 mmol/g loading

matrix

crosslinked with 2% DVB

particle size

200-400 mesh

storage temp.

2-8°C

Other Notes

Polymer-bound reagent for introducing the Alloc protecting group

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Dendrinos et al.
Journal of the Chemical Society. Perkin Transactions 1, 1463 -1463 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service