Skip to Content
Merck
All Photos(1)

Key Documents

675660

Sigma-Aldrich

2-(Tributylstannyl)pyrazine

95%

Synonym(s):

(2-Pyrazinyl)tributyltin, Tributylstannylpyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H3N2)Sn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
369.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

refractive index

n20/D 1.5158

bp

130 °C/1 mmHg

density

1.1706 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1cnccn1

InChI

1S/C4H3N2.3C4H9.Sn/c1-2-6-4-3-5-1;3*1-3-4-2;/h1-3H;3*1,3-4H2,2H3;

InChI key

OVBXTKIWZAHFAC-UHFFFAOYSA-N

Application

Microwave-assisted synthesis of substituted pyranones by palladium-catalyzed cross-coupling of arylstannanes with pyranyl triflates.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 48, 2063-2063 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service