650900
2-Bromo-1-(p-toluenesulfonyl)pyrrole
90%, technical grade
About This Item
grade
technical grade
Assay
90%
impurities
≤11% 1-(p-tolylsulfonyl)pyrrole
mp
102-105 °C (lit.)
SMILES string
Cc1ccc(cc1)S(=O)(=O)n2cccc2Br
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service