Skip to Content
Merck
All Photos(1)

Key Documents

596477

Sigma-Aldrich

4-(Triethoxysilyl)aniline

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3SiC6H4NH2
CAS Number:
Molecular Weight:
255.39
MDL number:
UNSPSC Code:
12352103

Assay

95%

bp

107-109 °C/0.1 mmHg (lit.)

density

1.502 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)c1ccc(N)cc1

Application

A silicon-based nucleophile shown to be reactive in Pd-catalyzed cross-coupling reactions.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denmark, S. E.; Ober, M. H.
Aldrichimica Acta, 36, 75-75 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service