Skip to Content
Merck
All Photos(1)

Documents

513938

Sigma-Aldrich

N-(2-Fluorophenyl)methanesulfonamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2NHC6H4F
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

75-77 °C (lit.)

SMILES string

CS(=O)(=O)Nc1ccccc1F

InChI

1S/C7H8FNO2S/c1-12(10,11)9-7-5-3-2-4-6(7)8/h2-5,9H,1H3

InChI key

DJLCKPRURZWUOU-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service