Skip to Content
Merck
All Photos(1)

Key Documents

446335

Sigma-Aldrich

Diethylmethylvinylsilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(C2H5)2CH=CH2
CAS Number:
Molecular Weight:
128.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.422 (lit.)

bp

118 °C (lit.)

density

0.75 g/mL at 25 °C (lit.)

SMILES string

CC[Si](C)(CC)C=C

InChI

1S/C7H16Si/c1-5-8(4,6-2)7-3/h5H,1,6-7H2,2-4H3

InChI key

BAVNDESSHRPRRF-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service