Skip to Content
Merck
All Photos(1)

Key Documents

424064

Sigma-Aldrich

Sodium iodate

≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaIO3
CAS Number:
Molecular Weight:
197.89
EC Number:
MDL number:
UNSPSC Code:
12352302

Assay

≥99.5%

density

4.28 g/mL at 25 °C (lit.)

SMILES string

[Na+].[O-]I(=O)=O

InChI

1S/HIO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

WTCBONOLBHEDIL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luisa M Franco et al.
Investigative ophthalmology & visual science, 50(8), 4004-4010 (2009-04-03)
To correlate damage to the retinal pigment epithelium (RPE) with decreased visual function after the systemic administration of sodium iodate (NaIO(3)). Damage was produced in mice by injection of 15, 25, or 35 mg/kg NaIO(3). Visual function was assessed with
William S Redfern et al.
Journal of pharmacological and toxicological methods, 63(1), 102-114 (2010-07-14)
We have evaluated the ability of a semi-automated, optomotor reflex method to assess drug-induced visual dysfunction, in albino and pigmented rats and mice. Male Han Wistar (HW) and Long Evans (LE) rats and mice (CD-1 and C57BL/6) were tested in
Huiming Xia et al.
Experimental eye research, 93(6), 862-872 (2011-10-13)
Regenerative medicine holds the promise of restoring cells and tissues that are destroyed in human disease, including degenerative eye disorders. However, development of this approach in the eye has been limited by a lack of animal models that show robust
Anna Machalińska et al.
Current eye research, 36(7), 663-672 (2011-06-11)
Stem cell regeneration of damaged tissue has recently been reported in many different organs. Here, we investigated the mobilization of different stem/progenitor cell (SPC) populations into the peripheral blood (PB), their subsequent homing to the injured retina (IR) and contribution
M H Cao et al.
Journal of hazardous materials, 179(1-3), 1143-1146 (2010-04-21)
The photochemical decomposition of perfluorooctanoic acid (PFOA) in aqueous periodate (IO(4)(-)) was investigated under two types of low-pressure mercury lamps: one emits at 254nm light (UV light) and the other emits both 254 nm and 185 nm light (VUV light).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service