Skip to Content
Merck
All Photos(1)

Key Documents

418471

Sigma-Aldrich

Phenylmaleic anhydride

99%

Synonym(s):

3-Phenyl-2,5-furandione, 3-Phenylfuran-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6O3
CAS Number:
Molecular Weight:
174.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99%

mp

120-122 °C (lit.)

SMILES string

O=C1OC(=O)C(=C1)c2ccccc2

InChI

1S/C10H6O3/c11-9-6-8(10(12)13-9)7-4-2-1-3-5-7/h1-6H

InChI key

QZYCWJVSPFQUQC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Won Kim et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-17)
Styrene can be formed by the microbial metabolism of bacteria and fungi. In our previous study, styrene was determined as a spoilage marker of Fuji apples decayed by Penicillium expansum, which is responsible for postharvest diseases. In the present study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service