Skip to Content
Merck
All Photos(1)

Documents

334804

Sigma-Aldrich

10-DOXYL-stearic acid, free radical

Synonym(s):

2-(8-Carboxyoctyl)-4,4-dimethyl-2-octyl-3-oxazolidinyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43NO4
CAS Number:
Molecular Weight:
385.58

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service