32578
Diethyl trimethylsilyl phosphite
≥95.0% (GC)
Synonym(s):
Phosphorous acid diethyl trimethylsilylester
About This Item
Recommended Products
Quality Level
Assay
≥95.0% (GC)
refractive index
n20/D 1.413 (lit.)
n20/D 1.413
bp
58-59 °C/10 mmHg (lit.)
density
0.921 g/mL at 25 °C (lit.)
SMILES string
CCOP(OCC)O[Si](C)(C)C
InChI
1S/C7H19O3PSi/c1-6-8-11(9-7-2)10-12(3,4)5/h6-7H2,1-5H3
InChI key
ATRMOIYYLVRRBZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
91.4 °F - closed cup
Flash Point(C)
33 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service