Skip to Content
Merck
All Photos(1)

Key Documents

292095

Sigma-Aldrich

2,2,2-Trifluoroethyl trifluoroacetate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CO2CH2CF3
CAS Number:
Molecular Weight:
196.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

3.34 psi ( 20 °C)

Assay

≥95%

refractive index

n20/D 1.277 (lit.)

bp

55 °C (lit.)

density

1.464 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

FC(F)(F)COC(=O)C(F)(F)F

InChI

1S/C4H2F6O2/c5-3(6,7)1-12-2(11)4(8,9)10/h1H2

InChI key

ZKUJOCJJXCPCFS-UHFFFAOYSA-N

General description

The molecular structure of 2,2,2-trifluoroethyl trifluoroacetate has been determined in the gas-phase from electron-diffraction data supplemented by ab initio (MP2) and DFT calculations. Hydrogenation of 2,2,2-trifluoroethyl trifluoroacetate catalyzed by anionic tris-phosphine ruthenium hydride complex has been reported.

Application

2,2,2-Trifluoroethyl trifluoroacetate has been used in the preparation of dimethyl (3,3,3-trifluoro-2,2-dihydroxypropyl)phosphonate.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

32.0 °F - closed cup

Flash Point(C)

0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental and theoretical studies of the vibrations and structure of 2, 2, 2-trifluoroethyl trifluoroacetate, CF3CO2CH2CF3.
Defonsi Lestard ME, et al.
Journal of Molecular Structure, 917(2), 183-192 (2009)
A convenient synthesis of dimethyl (diazomethyl) phosphonate (Seyferth/Gilbert reagent).
Brown DG, et al.
The Journal of Organic Chemistry, 61(7), 2540-2541 (1996)
Homogeneous catalytic hydrogenation of carboxylic acid esters to alcohols.
Grey RA, et al.
Journal of the Chemical Society. Chemical Communications, 16, 783-784 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service