All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H3ClN4
CAS Number:
Molecular Weight:
154.56
Beilstein:
5774
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
≥95.0% (HPLC)
color
yellow-orange
mp
>300 °C (dec.) (lit.)
solubility
DMF: soluble 0.2 g/10 mL, clear, colorless to light greenish-yellow
SMILES string
Clc1ncnc2[nH]cnc12
InChI
1S/C5H3ClN4/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H,7,8,9,10)
InChI key
ZKBQDFAWXLTYKS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
The acid-catalyzed reaction of 6-chloropurine with 3,4-di-O-acetyl-D-xylal has been investigated.
Application
6-Chloropurine has been used in the preparation of:
- 9-alkylpurines via alkylation with various substituted alkyl halides in DMSO
- 6-succinoaminopurine
Quantity
λmax. 264 nm, mol. absorption 8700 (0.1 M HCl)
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of 6-succinoaminopurine.
C E CARTER
The Journal of biological chemistry, 223(1), 139-146 (1956-11-01)
Heterocyclic N-glycosides-V: Synthesis of unsaturated N-glycosides from 6-chloropurine and derivatives of d-xylal and l-arabinal. A conformational NMR study.
Fuertes M, et al.
Tetrahedron, 26(20), 4823-4837 (1970)
Synthesis of Potential Anticancer Agents. XXVI. The Alkylation of 6-Chloropurine2.
Montgomery JA and Temple Jr C.
Journal of the American Chemical Society, 83(3), 630-635 (1961)
Takayuki Ando et al.
Carbohydrate research, 342(17), 2641-2648 (2007-09-14)
1',2'-cis-beta-Glycosyladenine nucleosides, such as beta-altroside, beta-mannoside, and beta-idoside, were efficiently synthesized from the corresponding 1',2'-trans-beta-6-chloropurine derivatives, beta-glucoside, and beta-galactoside. Nucleophilic substitution of the O-trifluoromethanesulfonyl groups at the C-2' and/or 3' was carried out using tetrabutylammonium acetate or cesium acetate under
Shi Bai et al.
Magnetic resonance in chemistry : MRC, 48(1), 61-67 (2009-11-26)
The (15)N and (13)C chemical shifts of 6-(fluoro, chloro, bromo, and iodo)purine 2'-deoxynucleoside derivatives in deuterated chloroform were measured. The (15)N chemical shifts were determined by the (1)H-(15)N HMBC method, and complete (15)N chemical-shift assignments were made with the aid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service