Skip to Content
Merck
All Photos(1)

Key Documents

260959

Sigma-Aldrich

Phenyl-1,4-benzoquinone

98%

Synonym(s):

Phenyl-p-benzoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8O2
CAS Number:
Molecular Weight:
184.19
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

113-115 °C (lit.)

SMILES string

O=C1C=CC(=O)C(=C1)c2ccccc2

Packaging

Packaged in glass bottles

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Miao et al.
Chemico-biological interactions, 317, 108945-108945 (2020-01-15)
Liver fibrosis is a common pathological consequence of liver injury, which increases liver failure-related morbidity and mortality. Hence, anti-fibrotic treatment is urgently needed. Oxidative stress plays a pivotal role in the progression of liver fibrosis. Thus, targeting ROS may be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service