Skip to Content
Merck
All Photos(1)

Documents

179051

Sigma-Aldrich

trans-2-Phenyl-1-cyclohexanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H10OH
CAS Number:
Molecular Weight:
176.25
Beilstein:
3198908
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

152-155 °C/16 mmHg (lit.)

mp

53-55 °C (lit.)

SMILES string

O[C@@H]1CCCC[C@H]1c2ccccc2

InChI

1S/C12H16O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-3,6-7,11-13H,4-5,8-9H2/t11-,12+/m0/s1

InChI key

AAIBYZBZXNWTPP-NWDGAFQWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The enantioselective resolution of trans-2-phenyl-1-cyclohexanol by Candida rugosa lipase was studied.

Application

trans-2-Phenyl-1-cyclohexanol was used in the synthesis of chiral compounds with high purity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic resolution of chiral alcohols in bifunctional membrane exhibiting enzyme activity and enantioselective permeation.
Journal of Molecular Catalysis. B, Enzymatic, 24, 17-26 (2003)
A Sánchez et al.
Journal of biotechnology, 84(1), 1-12 (2000-10-18)
Enantioselective resolution of trans-2-phenyl-1-cyclohexanol (TPCH) by a Candida rugosa lipase, obtained by fermentation in the laboratory, and immobilised on EP100 polypropylene powder has been carried out using isooctane as solvent and propionic acid as esterifying agent. The study have included

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service