Skip to Content
Merck
All Photos(1)

Key Documents

1.46625

Millipore

FREY Liquid Medium

pkg of 800 mL (in 1 L plastic bottle with screw cap), suitable for sterility testing, selective for mycoplasma

Synonym(s):

FREY Broth, Mycoplasma Broth, Mycoplasma Liquid Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225

Pricing and availability is not currently available.

Agency

EP (2.6.7)

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

6 mo.

feature

ready-to-use

packaging

pkg of 800 mL (in 1 L plastic bottle with screw cap)

technique(s)

microbiological culture: suitable

pH

7.6-7.8

application(s)

industrial qc
pharmaceutical
pharmaceutical

sterility testing

General description

FREY liquid Medium is designed for qualitative detection of avian mycoplasmas. The medium is supplemented with NAD (Nucleotide Adenine Dinucleotide) to allow the growth of M. synoviae. Color change of the medium to yellow or purple indicates the growth of mycoplasmas, due to Phenol Red in the medium. Penicillin inhibits the growth of accompanying bacterial flora.

Application

FREY Broth is recommended for the detection of avian mycoplasmas in pharmaceutical products, especially in bulk vaccines, cell banks, and virus cultures.

Features and Benefits

  • Ready-to-use convenience as no further preparation steps are required.
  • Suitable for pharmaceutical products such as Biopharmaceuticals, Vaccines, Cell cultures, and Virus cultures.
  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy and reducing the risk of incorrect interpretation and false results.

Analysis Note

The appearance of the medium is clear and red and color change reaction from red to yellow determines the presence of mycoplasma.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service