Skip to Content
Merck
All Photos(1)

Key Documents

Z280453

Sigma-Aldrich

Quantofix® test strips

Nitrite (NO2-)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034
NACRES:
NB.35
Pricing and availability is not currently available.

description

range mg/L 0, 100, 300, 600, 1000, 2000, 3000
semi-quantitative

specific analyte(s)

nitrite

reaction suitability

reaction type: Griess reaction

packaging

pack of 100 ea

manufacturer/tradename

Macherey-Nagel

color

orange to red change

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

General description

For semi-quantitative determination of ions and other substances. Consists of plastic strip to which test paper has been sealed. Size of test paper area permits clear comparison with fields of equal size on color scale provided on plastic box. Length sufficient to permit easy handling, even when aggressive or otherwise noxious solutions are being tested.

  • Fast; dip and read
  • No apparatus needed, only single test stick for analysis
  • Highly specific; interferences can be eliminated by precipitation or masking
  • Dim.: 6 × 95 × 0.2 mm

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service