Skip to Content
Merck
Quantofix® test strips Iron 1000

Quantofix® test strips

Quantofix® test strips Iron 1000

Products

Product Category
Brand
Color
Manufacturer
Analyte Suitability
Detection Method
Application
Package Size
Reaction Type