ZRB2395
Anti-p-MST1/2 (Thr183/180) Antibody, clone 1C2 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
EC: 2.7.11.1, MST-1/2, STE20-like kinase MST1/2, Mammalian STE20-like protein kinase 1/2, STK4/3, Serine/threonine-protein kinase 4/3, Serine/threonine-protein kinase Krs-2/1
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1C2, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
purified by
using Protein A
species reactivity
human
species reactivity (predicted by homology)
rat, mouse
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal half
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... STK4/STK3(6789)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in lysate from HeLa cells treated with Staurosporine.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-MST1 (Thr183)/MST2 (Thr180) in lysate from overnight serum starved HeLa cells treated Staurosporine (0.5 M; 6 h), but not in lysate from untreated cells.
Tested Applications
Affinity Binding Assay: A representative lot of this antibody bound phosphorylated MST1/MST2 peptide ((Thr183/180) with a KD of 4.4 x 10-6 in an affinity binding assay.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected p-MST1/2 (Thr183/180) in HeLa cells treated with Staurosporine.
Peptide Inhibition Assay:: Target band detection in HeLa cell lysate was prevented by pre-blocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service