Skip to Content
Merck
All Photos(8)

Key Documents

HPA021037

Sigma-Aldrich

Anti-ALDH1B1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Aldehyde dehydrogenase 5, Anti-Aldehyde dehydrogenase X, mitochondrial, Anti-Aldehyde dehydrogenase family 1 member B1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€545.00

€545.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€545.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€545.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

MLRFLAPRLLSLQGRTARYSSAAALPSPILNPDIPYNQLFINNEWQDAVSKKTFPTVNPTTGEVIGHV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALDH1B1(219)

General description

The gene ALDH1B1 (aldehyde dehydrogenase family 1 member B1) is mapped to human chromosome 9p11.1. It belongs to ALDH superfamily of enzymes. The protein localizes in the mitochondria. ALDH1B1 is expressed in liver, stomach and small intestine. The protein exists as a homotetramer.

Immunogen

Aldehyde dehydrogenase X, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ALDH1B1 (aldehyde dehydrogenase family 1 member B1) is mainly responsible for metabolism of acetaldehyde and lipid peroxidation products. It also takes part in ethanol detoxification pathway. ALDH1B1 is linked with development of coronary artery disease. Single nucleotide polymorphism in ALDH1B1 is associated with drinking patterns in humans. It is up-regulated in colonic adenocarcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75290

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuefei Wang et al.
PloS one, 9(7), e103161-e103161 (2014-07-23)
Coronary artery disease (CAD) is a multifactorial and polygenic disease. The aim of this study was to examine the association between six polymorphisms of four alcohol metabolism relevant genes (ADH1B, ADH1C, ALDH1b1, ALDH2) and the risk of CAD in Han
Peter Bjerregaard et al.
Drug and alcohol dependence, 144, 239-244 (2014-10-15)
Variation in genes involved in alcohol metabolism is associated with drinking patterns worldwide. We compared variation in these genes among the Inuit with published results from the general population of Denmark and, due to the Asian ancestry of the Inuit
Ying Chen et al.
Biochemical and biophysical research communications, 405(2), 173-179 (2011-01-11)
Aldehyde dehydrogenases (ALDHs) belong to a superfamily of NAD(P)+-dependent enzymes, which catalyze the oxidation of endogenous and exogenous aldehydes to their corresponding acids. Increased expression and/or activity of ALDHs, particularly ALDH1A1, have been reported to occur in human cancers. It
Brian C Jackson et al.
Chemico-biological interactions, 202(1-3), 11-21 (2012-12-19)
Vertebrate ALDH2 genes encode mitochondrial enzymes capable of metabolizing acetaldehyde and other biological aldehydes in the body. Mammalian ALDH1B1, another mitochondrial enzyme sharing 72% identity with ALDH2, is also capable of metabolizing acetaldehyde but has a tissue distribution and pattern
Mariana Lopes dos Santos et al.
International journal of oncology, 28(6), 1441-1461 (2006-05-11)
Over-expression of ERBB2, a member of the family of transmembrane receptor tyrosine kinases, occurs in 15-30% of primary breast tumors and is associated with poor prognosis and chemoresistance to a variety of anticancer drugs. In this study, aiming to identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service