Skip to Content
Merck
All Photos(1)

Key Documents

CLS2506

Corning® DNA-BIND® 1 x 8 Stripwell 96 well plates

1 x 8 Stripwell; 96 well plate, DNA-BIND, polystyrene, flat bottom, non-sterile, clear, 50/cs

Synonym(s):

1 x 8 stripwell, 1 x 8 stripwell plate, 96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, stripwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
material:
clear bottom
clear wells
flat wells
polystyrene
round wells
size:
96 wells , 1 x 8 Stripwell
Product CLS2506 is not currently sold in your country. Contact Technical Service

material

clear bottom
clear wells
flat wells
polystyrene
round wells

sterility

non-sterile

feature

lid: no
skirt
8-well strips
plate format: 96 well Stripwell
well: flat bottom, clear

packaging

pack of 1
case of 50

manufacturer/tradename

Corning 2506

size

96 wells , 1 x 8 Stripwell

well volume

50 μL

well working volume

75-200 μL

color

clear

suitability

suitable for (hybridization, amplification and other DNA-based assays)

binding type

DNA-BIND® surface

Looking for similar products? Visit Product Comparison Guide

General description

These 1 x 8 Stripwell 96 well plates have flat bottom polystyrene strips that are designed to easily break apart and are pre-assembled in an "egg-crate" style strip holder that allows each individual well to be positioned back into the plate once broken. DNA-BIND surface covalently binds to amine groups, and provides a convenient method to covalently immobilize aminated ssDNA by either the 5′ or 3′ end for hybridization and/or amplification, and for other DNA-based assays.

Corning DNA-BIND 1 x 8 Stripwell plates have the following features:
  • Polystyrene strips
  • Flat well bottoms
  • Total well volume of 360 μL; working well volumes of 75 to 200 μL
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation
  • Suitable for hybridization, amplification and other DNA-based assays
  • Lids are not available for these microplates

Legal Information

Corning is a registered trademark of Corning, Inc.
DNA-BIND is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service