Skip to Content
Merck
All Photos(1)

Key Documents

CHIRP03

Sigma-Aldrich

EZ-Magna ChIRP® Components

Sign Into View Organizational & Contract Pricing

Select a Size

CHIRP03
€706.00

€706.00


Estimated to ship onSeptember 30, 2025Details


Request a Bulk Order

Select a Size

Change View
CHIRP03
€706.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

€706.00


Estimated to ship onSeptember 30, 2025Details


Request a Bulk Order

Quality Level

Legal Information

MAGNA CHIRP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service