Skip to Content
Merck
All Photos(2)

Key Documents

8.05922

Sigma-Aldrich

Manganese(II) acetylacetonate

for synthesis

Synonym(s):

Manganese(II) acetylacetonate, Bis(acetylacetonato)manganese(II), Bis(2,4-pentanedionato)manganese(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14MnO4
CAS Number:
Molecular Weight:
253.15
MDL number:
UNSPSC Code:
12352103
EC Index Number:
237-858-2

form

powder

solubility

11.5 g/L

bulk density

550 kg/m3

storage temp.

2-30°C

Analysis Note

Assay (out of Mn): ≥ 97.0 %
Identity (IR-spectrum): conforms

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service