Skip to Content
Merck
All Photos(1)

Key Documents

MAN375326488

Manchester Organics

3,3,4,4,4-Pentafluorobutan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
163.09
MDL number:
Pricing and availability is not currently available.

Iupac Name

3,3,4,4,4-pentafluoro-1-butanamine

Inchi Code

1S/C4H6F5N/c5-3(6,1-2-10)4(7,8)9/h1-2,10H2

InChI key

QPURPAZMWFFRHE-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/Y32268

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library