The maximum recommended operating temperature is 35°C.
Z249254
Quantofix® peroxides test sticks
range mg/L 0, 0.5, 2, 5, 10, 25
About This Item
Recommended Products
description
semi-quantitative
specific analyte(s)
hydrogen peroxide
reaction suitability
reaction type: Redox Reactions
packaging
pkg of 100 ea
manufacturer/tradename
Macherey-Nagel
detection method
colorimetric
Looking for similar products? Visit Product Comparison Guide
General description
An aqueous medium is needed.
Method of application:
Remove only as many test sticks as are required, and reseal the container immediately after use. Do not touch the test paper zone. Dip test stick briefly (approx. 1 second) into the test solution. After 5 seconds, compare the test paper zone with the color scale.
In the presence of hydrogen peroxide the test paper turns blue. QUANTOFIX®-Peroxide test sticks can also be used for the determination of peracetic acid and other organic and inorganic hydroperoxides. To test for hydroperoxides in organic solvents the test zone is wetted with one drop of water after evaporation of the solvent.
Interferences:
In the pH range of 2-9 the accuracy of the determination is independent of the pH of the test solution. Buffer strongly acidic solutions with sodium acetate and adjust alkaline solutions to about pH 5-7 with citric acid. Falsely positive results can only be caused by strong oxidizing agents.
Storage:
Avoid exposing the sticks to sunlight and moisture. Store unopened packs in refrigerator (+2°C to +8°C). Opened packs: store container in a cool and dry place.
An aqueous medium is needed.
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the maximum operating temperature for Quantofix® peroxides test sticks, Product Z249254?
1 answer-
Helpful?
-
-
What is the accuracy of the Quantofix® peroxides test sticks, Product Z249254?
1 answer-
The results obtained from the test sticks are considered to be qualitative, not quantitative.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
How do I use the Quantofix® peroxides test sticks, Product Z249254?
1 answer-
Remove only as many test sticks as required and reseal the container immediately after use. Do not touch the test paper zone. Dip the test stick briefly (approximately 1 second) into the test solution. After 20 seconds, compare the test paper zone with the color scale.
Helpful?
-
-
What color or colors will the Quantofix® peroxides test sticks, Product Z249254, turn in the presence of peroxides?
1 answer-
The indicating portion of the test stick will turn from white or off-white to a shade of blue or purple. That color may be correlated with the colors on the "chips" printed on the label of the container.
Helpful?
-
-
How should the Quantofix® peroxides test sticks, Product Z249254, be stored?
1 answer-
Avoid exposing the stocks to sunlight and moisture. Store the container below 30°C in a dry place.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service