Merck
主页药物递送埃洛石纳米管在纳米材料研究中的应用
埃洛石纳米管在纳米材料研究中的应用
Prof. Yuri Lvov, Dr. Ronald Price
Institute for Micromanufacturing, Louisiana Tech University, Ruston, LA 71272
埃洛石是一种天然存在的硅铝酸盐纳米管,其已被人们所遗忘。埃洛石(Al2Si2O5(OH)4•2H2O)是一种两层硅铝酸盐,主要为亚微米范围的中空管状结构,化学性质类似于高岭土1,2。相邻的氧化铝和二氧化硅层及其水合水,由于填充混乱可通过弯曲并形成多层管(图1)。埃洛石是一种经济的材料,可以作为原始矿物从相应的矿床中开采获得。2 对于大多数天然材料,埃洛石颗粒的大小在1-15微米长和10-150 nm内径之间变化,具体取决于矿床。在我们的研究中,我们使用了Applied Minerals公司生产的埃洛石G(目录号:685445)该材料的平均管直径为50 nm,内腔直径为15 nm(图1)。该埃洛石的典型比表面积为65 m2/g;孔体积约为1.25 mL/g;折射率为1.54;比重为2.53 g/cm3。 
埃洛石纳米管图像
图 1.直径为50 nm的埃洛石纳米管685445(a)和管截面(b)的TEM图像。
化学上,埃洛石纳米管的外表面具有类似于SiO2的特性,而内筒芯与Al2O3相似。埃洛石颗粒的电荷(ζ电位)行为可以粗略描述为主要具有负表面电势(pH为6-7)的SiO2的叠加,同时加上少部分带正电的Al2O3内表面。3,4 内腔的正电荷(低于pH 8.5)可增加埃洛石纳米管对负离子大分子的负载,而这些大分子同时被带负电荷的外表面排斥。各种活性剂,包括药物、5-9 烟酰胺腺嘌呤二核苷酸(NAD)、以及海洋生物杀虫剂,9 可包埋至其内腔以及多层铝硅酸盐壳的空隙中。经过包封之后可以再对药剂进行保留和释放,这使得埃洛石成为一种非常适合大分子递送应用的纳米材料。
我们对将大分子从埃洛石纳米管中进行装载和释放进行了研究。装载的操作如下:将包含埃洛石和目标分子饱和溶液的混合物的小瓶转移到真空罐中,然后使用真空泵反复抽真空。悬浮液轻微起泡,表明空气已从埃洛石内部除去。停止起泡后,将小瓶密封30分钟以达到分子分布平衡。将埃洛石悬浮液离心以除去过量的溶解分子并洗涤。该过程可以重复两次,以确保埃洛石被最大量的目标分子填充。通过这种简单的装载技术,研究人员可以类似于往枪口装子弹的形式,向纳米管腔中填充任何材料,例如药物、蛋白质、聚电解质、以及具有合适直径的纳米粒子。
在对埃洛石表面进行适当的预处理后,疏水剂和亲水剂都可以被截留。10-11 例如,我们最近证明了尿素酶负载的埃洛石可用于在管腔内进行生物催化CaCO3的合成。11 对于亲水性分子(例如kellin、NAD、四环素)来说,典型的释放时间为2-5小时;对于在水中溶解度较低的分子(例如地塞米松、呋喃苯胺酸、硝苯地平)来说,释放时间为5-20小时(图2)。通过纳米管腔开口的一维扩散模型可以很好的对从埃洛石中释放大分子的过程进行描述。
从埃洛石纳米管中释放大分子
从埃洛石纳米管中释放大分子
图 2.从埃洛石纳米管释放大分子(NAD、地塞米松、呋喃苯胺酸、硝苯地平)。陡峭的曲线表示分子微晶的直接溶解而无管负载。
我们可以预见大量的基础研究以及应用将会采用埃洛石粘土纳米管。研究将包括:
  • 通过制造塞子或缩小纳米管腔的出口来控制大分子的装载和释放
  • 通过将装载的分子和溶液中的分子在埃洛石纳米管的开口处进行反应,以发展出微管纳米反应器的概念
埃洛石纳米管的可能应用包括:
  • 防腐剂的控制释放
  • 除草剂、杀虫剂、杀真菌剂和抗微生物剂的持续释放
  • 药物、食品添加剂和香料的持续释放
  • 棒状纳米颗粒的模板合成
  • 用作催化载体和分子筛
  • 特异性离子吸附
  • 用作塑料填充料以增强强度和防止划伤
  • 用于高级陶瓷材料,尤其是生物相容性植入物。
材料
抱歉,发生了意外错误
Network error: Failed to fetch
1.
Joussein E, Petit S, Churchman J, Theng B, Righi D, Delvaux B. 2005. Halloysite clay minerals ? a review. Clay miner.. 40(4):383-426. https://doi.org/10.1180/0009855054040180
2.
Hope EW, Kittrick JA. 1964. Surface tension and the morphology of halloysite. Am. Mineral. 49(7):859-63.
3.
Tar?? G, Bobos I, Gomes CS, Ferreira JM. 1999. Modification of Surface Charge Properties during Kaolinite to Halloysite-7Å Transformation. Journal of Colloid and Interface Science. 210(2):360-366. https://doi.org/10.1006/jcis.1998.5917
4.
Baral S, Brandow S, Gaber BP. 1993. Electroless metalization of halloysite, a hollow cylindrical 1:1 aluminosilicate of submicron diameter. Chem. Mater.. 5(9):1227-1232. https://doi.org/10.1021/cm00033a008
5.
Levis S, Deasy P. 2003. Use of coated microtubular halloysite for the sustained release of diltiazem hydrochloride and propranolol hydrochloride. International Journal of Pharmaceutics. 253(1-2):145-157. https://doi.org/10.1016/s0378-5173(02)00702-0
6.
2001. Contributed Papers in Specimen Mineralogy: 27th Rochester Mineralogical Symposium. Rocks & Minerals. 76(4):252-258. https://doi.org/10.1080/00357520109603226
7.
Yang L, Fassihi R. 1997. Examination of drug solubility, polymer types, hydrodynamics and loading dose on drug release behavior from a triple-layer asymmetric configuration delivery system. International Journal of Pharmaceutics. 155(2):219-229. https://doi.org/10.1016/s0378-5173(97)00164-6
8.
Lvov Y, Price R, Gaber B, Ichinose I. 2002. Thin film nanofabrication via layer-by-layer adsorption of tubule halloysite, spherical silica, proteins and polycations. Colloids and Surfaces A: Physicochemical and Engineering Aspects. 198-200375-382. https://doi.org/10.1016/s0927-7757(01)00970-0
9.
R. Price, B. P. Gaber, Y. Lvov R. 2001. In-vitro release characteristics of tetracycline HCl, khellin and nicotinamide adenine dineculeotide from halloysite; a cylindrical mineral. Journal of Microencapsulation. 18(6):713-722. https://doi.org/10.1080/02652040010019532
10.
Lvov YM, Shchukin DG, Möhwald H, Price RR. 2008. Halloysite Clay Nanotubes for Controlled Release of Protective Agents. ACS Nano. 2(5):814-820. https://doi.org/10.1021/nn800259q
11.
Shchukin D, Sukhorukov G, Price R, Lvov Y. 2005. Halloysite Nanotubes as Biomimetic Nanoreactors. Small. 1(5):510-513. https://doi.org/10.1002/smll.200400120