Merck
LC/MS Analysis of Methadone and Metabolites EDDP and EMDP on Ascentis® Express RP Amide
LC/MS Analysis of Methadone and Metabolites EDDP and EMDP on Ascentis® Express RP Amide application for HPLC
材料
分析色谱柱
产品编号
说明
价格
Ascentis® Express RP-Amide 2.7μm HPLC 色谱柱
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
标准
产品编号
说明
价格
(+/-)-美沙酮标准液 溶液
1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
EDDP高氯酸盐标准液 CRM 溶液
1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®
EMDP盐酸盐标准液 盐酸盐 溶液
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express RP Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase
[A] 10 mM ammonium formate in water, pH 3.6; [B] 10 mM ammonium formate in acetonitrile, pH 3.6; (65:35, A:B)
flow rate
0.5 mL/min
column temp.
35 °C
detector
ESI(+), m/z 100-1000
injection
0.5 μL
sample
200 μg/L in 25:75, water: acetonitrile