Merck
HPLC Analysis of Peptides on Discovery® BIO Wide Pore C5
HPLC Analysis of Peptides on Discovery® BIO Wide Pore C5 application for HPLC
材料
分析色谱柱
产品编号
说明
价格
Discovery® BIO Wide Pore C5 高效液相色谱柱
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery BIO Wide Pore C5, 15 cm × 4.6 mm I.D., 5 μm particles (568422-U)
mobile phase
[A]: 0.1% PFPA (pentafluoropropionic acid) in water:acetonitrile (81:19); [B]: 0.1% PFPA in water:acetonitrile (62:38)
gradient
0-100% B in 19 min
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 215 nm
injection
10 μL ~0.25 μg each peptide
sample
Cat. No. P2693
说明
法律信息
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany