Merck
HPLC Analysis of Barbiturates on Discovery® C18
HPLC Analysis of Barbiturates on Discovery® C18 application for HPLC
材料
分析色谱柱
产品编号
说明
价格
SUPELCOSIL LC-ABZ HPLC 色谱柱
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] acetonitrile; [B] 25 mM potassium phosphate buffer, pH 6.9 (25:75, A:B)
flow rate
1.5 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
25 μL
说明
法律信息
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany