跳转至内容
Merck
  • [Potentiometric glucose determination in human serum samples with glucose oxidase biosensor based on iodide electrode].
[Potentiometric glucose determination in human serum samples with glucose oxidase biosensor based on iodide electrode].
Prikladnaia biokhimiia i mikrobiologiia (2013-06-26)
E Karakus, Sh Pekyardimchi, E Kilich
摘要
Glucose potentiometric biosensor was prepared by immobilizing glucose oxidase on iodide-selective electrode. The hydrogen peroxide formed after the oxidation of glucose catalysed by glucose oxidase (GOD) was oxidized by sodium molybdate (SMo) at iodide electrode in the presence ofdichlorometane. The glucose concentration was calculated from the decrease of iodide concentration determined by iodide-selective sensor. The sensitivity of glucose biosensor towards iodide ions and glucose was in the concentration ranges of 1.0 x 10(-1) - 1.0 x 10(-6) M and 1.0 x 10(-2) - 1.0 x 10(-4) M, respectively. The characterization of proposed glucose biosensor and glucose assay in human serum were also investigated.
材料
货号
品牌
产品描述
Sigma-Aldrich
二氯甲烷, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
二氯甲烷, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
二氯甲烷, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
钼酸钠 二水合物, ≥99.5%
Sigma-Aldrich
二氯甲烷, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
二氯甲烷, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
钼酸钠 二水合物, ACS reagent, ≥99%
Sigma-Aldrich
钼酸钠, ≥98%
Sigma-Aldrich
过氧化氢 溶液, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
二氯甲烷, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
二氯甲烷, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
钼酸钠 二水合物, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
二氯甲烷, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
二氯甲烷, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Millipore
过氧化氢 溶液, 3%, suitable for microbiology
Sigma-Aldrich
钼酸钠, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
钼酸钠 二水合物, 99.99% trace metals basis
Sigma-Aldrich
钼, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
钼, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
钼, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Supelco
二氯甲烷, analytical standard
Sigma-Aldrich
钼, wire, diam. 1.0 mm, 99.95% trace metals basis
Supelco
二氯甲烷, Selectophore, ≥99.5%
Sigma-Aldrich
二氯甲烷, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
钼, foil, thickness 1.0 mm, ≥99.9% trace metals basis