主要文件
712-0266
GC Analysis of Lactone Enantiomers on β-DEX 110
suitable for GC
登录查看公司和协议定价
About This Item
技術
gas chromatography (GC): suitable
測試參數
column: β-DEX 110, 30 m × 0.25 mm I.D., 0.25 μm (24301)
oven: 90 °C, 1 °C/min to 200 °C
inj. temp.: 300 °C
detector: FID, 300 °C
carrier gas: helium, 20 cm/sec @ 90 °C
injection: 1 μL, 100:1 split
sample: 0.5 mg/mL in methylene chloride
應用
chemicals and industrial polymers
flavors and fragrances
法律資訊
DEX is a trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)