主要文件
509744
用于 PerkinElmer® Autosystem 气相色谱仪的 SGE 注射器
volume 5 μL, 5F-PE-0.63, needle size 23 ga, needle L 70 mm, needle type, LTN (fixed)
登录查看公司和协议定价
选择尺寸
1 EA
$187.00
$187.00
请联系客服了解存货情况
获取大包装报价
选择尺寸
变更视图
1 EA
$187.00
About This Item
UNSPSC代码:
41122001
$187.00
请联系客服了解存货情况
获取大包装报价
物料
cone tip stainless steel needle (point style 1)
针尺寸
23 ga
描述
liquid syringe, fixed needle
特点
needle type LTN (fixed)
包装
pkg of 1 ea
制造商/商品名称
SGE 001954
针长度
70 mm
体积
5 μL
正在寻找类似产品? 访问 产品对比指南
一般描述
SGE Syringes for PerkinElmer® Autosystem [volume 5 μL, 5F-PE-0.63, needle size 23 ga (cone tip), needle L 70 mm, needle type, LTN (fixed)] are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most trouble-free autosampler syringes available. The stainless steel cone tip, point style #1 is recommended for use with pre-drilled septa. The shape of this needle on the syringe has been developed for multiple and fast septum injections on the autosampler.
法律信息
PerkinElmer is a registered trademark of PerkinElmer Inc.
历史批次信息供参考:
分析证书(COA)