主要文件
Z757365
Sartorius Proline® Plus mechanical pippettor
single channel, volume 5-50 μL
登录查看公司和协议定价
选择尺寸
1 EA
$442.00
$442.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$442.00
About This Item
UNSPSC代码:
41121500
NACRES:
NB.01
$442.00
请联系客服了解存货情况
特点
electronic: no
single-channel
包装
pack of 1 ea
制造商/商品名称
Sartorius 728040
体积
5-50 μL
正在寻找类似产品? 访问 产品对比指南
一般描述
The Sartorius Proline Plus pipettes of the Biohitfamily are high quality with a light pipetting force. With robust design, the Proline Plus is suitable for variable every day laboratory applications and covers the full range of 0.1μL to 10 mL.
  • Reduced pipetting forces prevent RSI and improve the results
  • Accurate and precise
  • Contamination free pipetting with Safe-Cone Filters
  • Robust design
  • Easiest pipette to maintain
法律信息
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
历史批次信息供参考:
分析证书(COA)