主要文件
Z740434
HistoTainer II prefilled specimen containers
M961, volume 120 mL
登录查看公司和协议定价
选择尺寸
96 EA
$132.00
$132.00
请联系客服了解存货情况
选择尺寸
变更视图
96 EA
$132.00
About This Item
UNSPSC代码:
41121800
NACRES:
NB.22
$132.00
请联系客服了解存货情况
物料
polyethylene cap
polypropylene container
包装
pack of 96 ea
制造商/商品名称
Simport M961-120FMA
体积
120 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
NON Tamper Evident Prefilled Specimen Containers, 50% filled with 10% Neutral Buffered Formalin
Especially designed for collection, transport and storage of histology specimens, Simport offers shatter resistant polypropylene containers, eliminating most problems of leakage and evaporation. Containers are manufactured from virgin, translucent polypropylene. The white lids are ribbed for easy opening when hands are wet or gloved while the jars are stackable for easy, safe storage. Jars are translucent and specimens can be viewed without having to open the lid. Closures are manufactured from virgin polyethylene with a unique integrated leak-resistant seal.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All materials used in manufacturing are free from latex. Containers are
95 kPa compliant. Available in many sizes from 20 to 120 ml. Packaging is offered in trays of 24, cases of 96.
The HistoTainer II is half filled with a choice of fixatives. 10% Neutral Buffered Formalin penetrates quickly, but fixes slowly. The Formalin is enhanced by a buffering capacity optimizing histological results by light microscopy and immunohistochemistry. Bouin, Hollande and Zinc Formalin fixatives are also available.
法律信息
HistoTainer is a trademark of Simport
历史批次信息供参考:
分析证书(COA)