主要文件
Z740384
Eco-Friendly SpecTainer, biodegradable
size 90 mL, sterile
登录查看公司和协议定价
选择尺寸
400 EA
$419.00
$419.00
预计发货时间2025年5月28日详情
选择尺寸
变更视图
400 EA
$419.00
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.22
$419.00
预计发货时间2025年5月28日详情
材料
polyethylene cap
polypropylene container
無菌
sterile
包裝
pack of 400 ea
製造商/商標名
Simport C567-90AQSECO
尺寸
90 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
The Simport Eco-Friendly SpecTainer will biodegrade to become some of the soil′s organic components in less than 7 years in constant contact with the soil instead of up to 400 years when using conventional plastics.The Simport Eco-Friendly SpecTainer, one of the most innovative specimen containers on the market today, is now available in a new version: The Eco-Friendly SpecTainer, a rapidly degradable Specimen Container. Plastics take hundreds of years to degrade naturally in the environment. The only condition that is necessary for the Eco-Friendly SpecTainer to biodegrade is constant contact with other degrading material. No heat, physical stress, oxygen or sunlight necessary. The Eco-Friendly SpecTainer has the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment.Does not contain heavy metals. No negative effect to its physical properties. No heat, light or mechanical stress needed for product breakdown. No toxic residue. Fully degradable within 7 years, aerobically or anaerobically.
法律資訊
SpecTainer is a trademark of Simport
历史批次信息供参考:
分析证书(COA)