主要文件
Z637327
Pursept®-A disinfectant replacement atomizer for 1L bottles
登录查看公司和协议定价
选择尺寸
1 L
$31.00
$31.00
请联系客服了解存货情况
选择尺寸
变更视图
1 L
$31.00
About This Item
UNSPSC代码:
41103200
NACRES:
NB.35
$31.00
请联系客服了解存货情况
制造商/商品名称
BRAND 44832
一般描述
The Pursept®-A disinfectant replacement atomizer is designed specifically for 1L bottles, providing an efficient and convenient solution for disinfecting surfaces in various settings. This atomizer ensures an even and controlled application of disinfectant, enhancing coverage and effectiveness.
其他说明
BR44832 (India)
法律信息
Pursept is a registered trademark of Merz GmbH & Co. KG
历史批次信息供参考:
分析证书(COA)