主要文件
Z332216
Bottle adapters for BRAND® Dispensers and Burettes
GL 32 thread, GL 28 bottle, polypropylene
登录查看公司和协议定价
选择尺寸
1 EA
$21.00
$21.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$21.00
About This Item
UNSPSC代码:
41121815
NACRES:
NB.01
$21.00
请联系客服了解存货情况
物料
polypropylene
制造商/商品名称
BRAND 704328
配件
GL 28 bottle
GL 32 thread
正在寻找类似产品? 访问 产品对比指南
一般描述
For all Dispensette® models as well as seripettor® and QuikSip
其他说明
BR704328 (India)
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)