主要文件
Z329827
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Distilled water, with venting valve (metal ball)
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103209
NACRES:
NB.17
物料
low-density polyethylene bottle
natural bottle
round bottle
white closure
wide-mouth bottle
特点
label Distilled water
with cap
with venting valve (metal ball)
包装
pack of 5 ea
制造商/商品名称
BRAND 144018
容量
250 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
其他说明
BR144018 (India)
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
配件
历史批次信息供参考:
分析证书(COA)