主要文件
Z328723
BRAND®磁性搅棒,PTFE,圆柱形
L 15 mm, bar diam. 6 mm
登录查看公司和协议定价
选择尺寸
10 EA
$60.50
$60.50
请联系客服了解存货情况
选择尺寸
变更视图
10 EA
$60.50
About This Item
UNSPSC代码:
41103806
eCl@ss:
32100100
NACRES:
NB.72
$60.50
请联系客服了解存货情况
物料
PTFE-coated
cylindrical
制造商/商品名称
BRAND 137114
长度
15 mm
柱直径
6 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
特点和优势
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
其他说明
BR137114 (India)
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)