主要文件
Z327069
BRAND® BLAUBRAND® volumetric flask, glass stopper, amber glass
volume 100 mL, accuracy: 0.1 mL
登录查看公司和协议定价
选择尺寸
2 EA
$134.00
$134.00
请联系客服了解存货情况
选择尺寸
变更视图
2 EA
$134.00
About This Item
UNSPSC代码:
41121804
NACRES:
NB.43
$134.00
请联系客服了解存货情况
物料
borosilicate glass
接合处
joint ST/NS neck14/23
无菌性
non-sterile
特点
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
closure type Glass stopper
包装
pack of 2
制造商/商品名称
BRAND
体积测量范围
100 mL, accuracy: 0.1 mL
容量
100 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from DURAN®, calibrated to contain (TC, IN). Supplied with a interchangeable amber glass stopper. Marks and inscriptions in high contrast white enamel.
All BLAUBRAND volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
其他说明
BR37449 (India)
法律信息
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)