主要文件
Z325562
BRAND® SILBERBRAND ETERNA graduated pipette, calibrated to deliver (TD, EX)
capacity 25 mL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
材料
AR-Glass
描述
includes enlarged suction tube for cotton plugs.
特點
class B
graduation
subdivision 0.1 mL
supplementary to DIN range
包裝
pack of 6
製造商/商標名
BRAND 27079
容量
25 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
AR-glass®. Graduated pipettes according to DIN EN ISO 835. Type 3 (zero point at the top), total delivery. Graduations and inscriptions in ETERNA amber stain, particularly resistant to aggressive cleaning methods.
其他說明
BR27079 (India)
法律資訊
AR-Glas is a registered trademark of Schott AG
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)
访问文档库
实验方案
This page contains detailed information on working with and handling pipettes and pipetting aids.
本页面包含操作移液管和助吸器的详细信息。