主要文件
TMO2116-0250
Nalgene® straight-sided wide-mouth jar
polycarbonate, jar capacity 250 mL, clear, case of 24 ea
别名:
clear plastic container, screw top container
登录查看公司和协议定价
选择尺寸
24 EACH
$574.00
$574.00
预计发货时间2025年6月04日详情
选择尺寸
变更视图
24 EACH
$574.00
About This Item
UNSPSC代码:
41121800
NACRES:
NB.43
$574.00
预计发货时间2025年6月04日详情
物料
clear
polycarbonate
straight
wide-mouth
包装
case of 24 ea
制造商/商品名称
Nalgene 2116-0250
罐容量
250 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Collect and store pastes, solids, specimens and lab supplies with window-clear Thermo Scientific Nalgene Straight-side Wide-Mouth Jars. A wide temperature range (-135°C to +135°C) makes these jars an excellent choice for cold room and refrigerated use all the way to the autoclave. These high impact-resistant jars are extremely versatile. Not liquid leakproof.
法律信息
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
历史批次信息供参考:
分析证书(COA)