主要文件
RFA4530113-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-30-11, aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103909
产品RFA4530113-EP监管中,请联系销售询价或购买 联系客户支持
特点
lid
to hold 30 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor
包装
pack of 1 ea
制造商/商品名称
Eppendorf® 022654047 (NA)
Eppendorf® 5427753001 (EU)
参数
20,817 × g max. RCF
技术
centrifugation: suitable
相容性
for use with centrifuge models 5430/5430R
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
特点和优势
  • The rotor features aerosol tight quick lock rotor lid made of aluminum for use with hazardous sample
  • Fixed angle rotor holds sample at 45°
  • Compatible with 5430/5430R centrifuges
法律信息
Eppendorf is a registered trademark of Eppendorf AG
历史批次信息供参考:
分析证书(COA)