主要文件
HS15981C
Slide holder
for 15 slides, white polyethylene
登录查看公司和协议定价
选择尺寸
1 EACH
$61.10
$61.10
预计发货时间2025年5月29日详情
选择尺寸
变更视图
1 EACH
$61.10
About This Item
UNSPSC代码:
41123302
NACRES:
NB.35
$61.10
预计发货时间2025年5月29日详情
物料
white polyethylene
特点
Holds 15 x slides 76x26 mm
制造商/商品名称
Heathrow Scientific HS15981C
尺寸
153x102x21 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
The Slide Holder holds slides during evaluation, cleaning, and drying.
  • Made from chemical-resistant polyethylene
  • Solid, strong construction with rubber feet for bench top stability
历史批次信息供参考:
分析证书(COA)