主要文件
HS120116
Nitrogen-free weighing papers
W × L 3 × 3 in.
登录查看公司和协议定价
选择尺寸
500 EACH
$65.00
$65.00
国内现货,预计发货时间2025年4月17日详情
选择尺寸
变更视图
500 EACH
$65.00
About This Item
UNSPSC代码:
41121812
NACRES:
NB.35
$65.00
国内现货,预计发货时间2025年4月17日详情
制造商/商品名称
Heathrow Scientific HS120116
宽度 × 长度
3 × 3 in.
颜色
white
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Nitrogen-free for a very smooth surface and good sample recovery. Strong enough to support dense material.
历史批次信息供参考:
分析证书(COA)
访问文档库
Questions
  1. What is the thickness of this butter paper?
    1 answer
    1. The thickness of this weighing paper is 0.0015” or 0.038 mm.